Orders LinkContactUsLinkComponent
Disposable Post Mortem scalpels
Disposable Post Mortem scalpels
PM40B, with bull nose blade
Catalogus # SWAN2581
Leverancier:   SWANN MORTON
Disposable Post Mortem scalpels
Catalogus # SWAN2581
Leverancier:  SWANN MORTON
Disposable Post Mortem scalpels
Disposable Post Mortem scalpels
PM40B, with bull nose blade
Price
122,00 €
Log in voor beschikbaarheid
1 * 5 ST
Product Details & Documents
Carbon blade. Non sterile, disposable histopathology and autopsy knives.
  • With easy to remove guard to protect the cutting edge: Users do not have to fix the blade to the handle
  • Available with either standard PM40 blade or PM40B bull nose blade fixed in a distinctive blue handle
Documents
Specificaties
  • Type:
    PM40B, with bull nose blade