Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
 

Petri Dishes

With 3 vents, 150 mm, 15 mm, Steriel, 11×20/bag
  ANIC00.50.18PAGI
 :   Anicrin
Petri Dishes
  ANIC00.50.18PAGI
 :  Anicrin
Petri Dishes
Petri Dishes

With 3 vents, 150 mm, 15 mm, Steriel, 11×20/bag

 
232,00 €
   
1 * 220 ST
 
  • Ø:
    150 mm
  • Verpakt:
    11×20/bag
  • Steriliteit:
    Steriel
  • Uitvoering:
    With 3 vents
  • Hoogte:
    15 mm
  • Seq:
    15