Bottles, water sampling
125 ml
Leverancier: LP ITALIANA
Ga naar:
Product Details & Documents
PP, translucent, square, with screw cap.
- Tamper evident screw caps
- Sterile
- Moulded graduations
Bottles for microbiological analysis of chlorinated or non-chlorinated water. Sodium thiosulphate inhibits the effects of chlorine, bromide and ozone present in the water. Ideal for flat storage (fridges, etc).
Documents
Specificaties
- Capaciteit:125 ml
- Seq:02