Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Contrast clips

Contrast clips

PP, 18,5 - 22,0 mm, 36 mm, wit
Catalog # WITG3.691.005
Supplier:   witeg Labortechnik
Contrast clips
Catalog # WITG3.691.005
Supplier:  witeg Labortechnik
Contrast clips
Contrast clips

PP, 18,5 - 22,0 mm, 36 mm, wit

Price
4,90 €
Log in for availability
1 * 1 ST
Product Details & Documents

PP, white/black.

  • For attachment to all measuring flasks
  • With grip deepening
  • Unbreakable, resistant to acids and alkalis
  • For better recognition and adjustment of the liquid level
Specifications
  • Spanwijdte:
    18,5 - 22,0 mm
  • Kleur:
    wit
  • Lengte:
    36 mm
  • Materiaal:
    PP
  • Seq:
    05