Contrast clips
PP, 10,0 - 13,3 mm, 36 mm, wit
WITG3.691.003
PP, white/black.
- For attachment to all measuring flasks
- With grip deepening
- Unbreakable, resistant to acids and alkalis
- For better recognition and adjustment of the liquid level
- Spanwijdte:10,0 - 13,3 mm
- Kleur:wit
- Lengte:36 mm
- Materiaal:PP
- Seq:03



