Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Contrast clips

Contrast clips

PP, 6,1 - 7,5 mm, 36 mm, wit
Catalog # WITG3.691.002
Supplier:   witeg Labortechnik
Contrast clips
Catalog # WITG3.691.002
Supplier:  witeg Labortechnik
Contrast clips
Contrast clips

PP, 6,1 - 7,5 mm, 36 mm, wit

Price
4,40 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents

PP, white/black.

  • For attachment to all measuring flasks
  • With grip deepening
  • Unbreakable, resistant to acids and alkalis
  • For better recognition and adjustment of the liquid level
Spécifications
  • Spanwijdte:
    6,1 - 7,5 mm
  • Kleur:
    wit
  • Lengte:
    36 mm
  • Materiaal:
    PP
  • Seq:
    02