Orders LinkContactUsLinkComponent
Verification standard kit, MultiDirect
Verification standard kit, MultiDirect
Verification standard kit, MD 100/MD200, 1
Catalogus # LOVI215670
Leverancier:   Lovibond
Verification standard kit, MultiDirect
Catalogus # LOVI215670
Leverancier:  Lovibond
CAS-nummer:  
Verification standard kit, MultiDirect
Verification standard kit, MultiDirect
Verification standard kit, MD 100/MD200, 1
CAS-nummer:  
Price
220,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Verification accuracy of photometers and validity of results.
  • Check the performance of photometers over the full measurement range
  • Results can be checked against tolerances provided on the certificate of conformity
Delivery information: Contains zero solution in sealed photometer vial, 6× verification standard solution in sealed photometer vial, light shield cap, certificate of conformity, instruction sheet.
Specificaties
Specificaties
  • VPE:
    1
  • Omschrijving:
    Verification standard kit, MD 100/MD200