Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Mop, AlphaMop™, PE

Mop, AlphaMop™, PE

Sterile AlphaMop™ polyester reserveovertrekken en kussens: STX7118, voor gebruik met TX7108 en TX7108A, 381×203,2 mm, -, 10 overtrekken en 1 kussen/zak; 12 zakken/doos
Catalogus # 141-1048
Leverancier:   Texwipe
Mop, AlphaMop™, PE
Catalogus # 141-1048
Leverancier:  Texwipe
Mop, AlphaMop™, PE
Mop, AlphaMop™, PE

Sterile AlphaMop™ polyester reserveovertrekken en kussens: STX7118, voor gebruik met TX7108 en TX7108A, 381×203,2 mm, -, 10 overtrekken en 1 kussen/zak; 12 zakken/doos

Price
847,54 €
Log in voor beschikbaarheid
1 * 1 KIT
Product Details & Documents

Ideal for cleaning ceilings, walls, floors, and other flat surfaces in critical environments. The changeable, laundered polyester head fits over a flat foam swivel pad assembly attached to a fibreglass handle. Replaceable foam pad ensures the polyester head conforms to textured floors and other surfaces while resisting corrosion and abrasion. A lightweight, durable fibreglass handle gives the user excellent control over the swivel head.

  • Lightweight mop maintains ultra-low generation of particles and extractables
  • Mop can be sterilised
  • Mophead covers can be pre-wetted with 6% isopropanol
  • Accesses hard-to-reach areas with ease
  • Excellent durability with low contamination
  • Allows user to change covers and foam pads frequently, adhering to wiping protocol
  • Rapid-change cover and foam pad configuration

* Handle length.

Verpakking: Mop is bag-within-a-bag® cleanroom packaged.

Let op: *The product(s) on this page are outside the scope of the EU Directive 2019/904.

Specificaties
  • Omschrijving:
    Sterile AlphaMop™ polyester reserveovertrekken en kussens: STX7118, voor gebruik met TX7108 en TX7108A
  • Lengte:
    -
  • l×b:
    381×203,2 mm
  • Seq:
    10
  • Verpakt:
    10 overtrekken en 1 kussen/zak; 12 zakken/doos
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.