Orders LinkContactUsLinkComponent
Conductivity standards
Conductivity standards
Sodium chloride standard solution, 491 mg/l NaCl, 1000 µS/cm, 500 mL
Catalogus # HACH1440049
Leverancier:   Hach
Conductivity standards
Catalogus # HACH1440049
Leverancier:  Hach
CAS-nummer:  
Conductivity standards
Conductivity standards
Sodium chloride standard solution, 491 mg/l NaCl, 1000 µS/cm, 500 mL
CAS-nummer:  
Price
74,70 €
Log in voor beschikbaarheid
1 * 500 mL
Product Details & Documents
The sodium chloride standard solutions are used for calibrating conductivity meters and also used for accuracy checks.
Specificaties
Specificaties
  • VPE:
    500 mL
  • Omschrijving:
    Sodium chloride standard solution, 491 mg/l NaCl
  • Geleidbaarheid:
    1000 µS/cm