Nessler cylinders
Nessler cylinder 100 ml, tall form, with optically plane bottom, circular mark at 100 ml, Ø×L: 32×220 mm
Catalogus # WITG2.150.100
Leverancier: witeg Labortechnik
Ga naar:
Product Details & Documents
Colour comparison cylinders according to Nessler for comparators, with optically plane bottom.
Specificaties
- Omschrijving:Nessler cylinder 100 ml, tall form, with optically plane bottom, circular mark at 100 ml, Ø×L: 32×220 mm
- Seq:02