Natriumhydroxide solution 50% (w/w), BAKER ANALYZED®, J.T.Baker®
IBC/tote, one way, 3.250 lb
Supplier: Avantor
Natriumhydroxide solution 50% (w/w), BAKER ANALYZED®, J.T.Baker®
Catalog # 3727-25
Supplier: Avantor
CAS Number: 1310-73-2
Jump to:
Product Details & Documents
MPORTANT: Material will freeze if stored below 60°F (16°C).
Caution: IMPORTANT: Material will freeze if stored below 60°F (16°C). Protect from air to avoid absorption of carbon dioxide.
Synonyms: Caustic soda
- CAS Number:1310-73-2
- Formula:NaOH
- MDL Number:MFCD00003548
- Molecular Weight:40 g/mol
- Boiling Pt:119…135 °C (1013 hPa)
- Melting Pt:9…10 °C
- Density:1,53 g/cm³ (20 °C)
- Storage Temperature:Kamertemperatuur
- EINECS:215-185-5
- UN:1824
- ADR:8,II
Documents
Certificates
Search by Lot NumberChemical Mining Supplies
Product Literature
sds_BAKR3727
SDS
spec_BAKR3727-03 spec_BAKR3727-09
Product specification
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:50 - 52%
- Protect from air to avoid absorption of carbon dioxide.:≤20
- Assay (NaOH) (by acidimetry):50 - 52 %
- Color (APHA):≤20
- Sodium Carbonate (Na₂CO₃):≤0.1 %
- Chloride (Cl):≤0.002 %
- Sulfate (SO₄):≤0.001 %
- Ammonium Hydroxide Precipitate:≤0.01 %
- Silica (SiO₂), For Information Only:≤5 ppm
- Heavy Metals (as Ag):≤0.001 %
- Potassium (K):≤0.01 %
- Nitrogen Compounds (as N):≤5 ppm
- Phosphate (PO₄):≤5 ppm
- Trace Impurities - Iron (Fe):≤5 ppm
- Trace Impurities - Nickel (Ni):≤5.000 ppm
Specifications
- Verpakking:IBC/tote, one way
- VPE:3.250 lb
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



