Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
 

Natriumhydroxide 25% in aqueous solution, J.T.Baker®

950 L
Catalog # 2613.9950
Corrosive
Supplier:   Avantor
Natriumhydroxide 25% in aqueous solution, J.T.Baker®
Catalog # 2613.9950
Supplier:  Avantor
CAS Number:  1310-73-2
Natriumhydroxide 25% in aqueous solution, J.T.Baker®

950 L

CAS Number:  1310-73-2
Price
Inquire For Price
Log in for availability
1 * 950 L
Product Details & Documents
Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Boiling Pt:119…135 °C (1013 hPa)
  • Melting Pt:9…10 °C
  • Density:1,53 g/cm³ (20 °C)
  • Storage Temperature:Kamertemperatuur
  • EINECS:215-185-5
  • UN:1824
  • ADR:8,II
Specifications

Specification Test Results

  • Assay (NaOH):
    24.4 - 25.6 %
  • Appearance (clear, colorless liquid):
    passes test
  • Endotoxin Concentration (EU/ml):
    max. 2.0
  • Sodium Carbonate (Na2CO3):
    max. 0.6 %
  • Trace Impurities (in ppm)Chlorides (as Cl):
    max. 5
  • Trace Impurities (in ppm)Iron (Fe):
    max. 0.5
  • Trace Impurities (in ppm)Heavy Metals (as Pb):
    max. 1

Specifications

  • VPE:
    950 L

Specifications