Orders LinkContactUsLinkComponent
qScript™ XLT One-Step RT-PCR Kit
qScript™ XLT One-Step RT-PCR Kit
qScript™ XLT One-Step RT-PCR Kit, 200 Assays
Catalogus # 733-2263
Leverancier:   Quantabio
qScript™ XLT One-Step RT-PCR Kit
Catalogus # 733-2263
Leverancier:  Quantabio
CAS-nummer:  
qScript™ XLT One-Step RT-PCR Kit
qScript™ XLT One-Step RT-PCR Kit
qScript™ XLT One-Step RT-PCR Kit, 200 Assays
CAS-nummer:  
Price
808,00 €
Log in voor beschikbaarheid
1 * 200 Essai
Product Details & Documents
The qScript™ XLT One-Step RT-PCR Kit is a convenient and highly sensitive system for amplification of RNA templates up to 2 kb. cDNA synthesis and PCR amplification are carried out in the same tube without opening between procedures. This system has been optimised to deliver maximum endpoint RT-PCR efficiency, sensitivity, and specificity.
  • Tough tested to overcome inhibitors
  • Option GelTrack™ Loading Dye included
  • Superior reproducibility
  • Convenient single tube reaction assembly at room temperature
Each kit comprises:
1×200 µl qScript™ XLT One-Step Reverse Transcriptase (25X concentrated mixture of qScript™ XLT reverse transcriptase and recombinant ribonuclease inhibitor protein)
2×1,25 ml One-Step ToughMix® (2X concentrated reaction buffer containing dNTP's, magnesium chloride, hot start DNA polymerase, and stabilisers)
1×0,4 ml GelTrack™ Loading Dye (50X concentrated mixture of RT-PCR compatible, blue and yellow electrophoresis tracking dyes)
2×1,5 ml nuclease-free water
Specificaties
Specificaties
  • VPE:
    200 Assays
  • Omschrijving:
    qScript™ XLT One-Step RT-PCR Kit
Specificaties