Hygromycin B
Glass bottle for solids, 100 mg
Leverancier: Thermo Fisher Scientific
Ga naar:
Product Details & Documents
Synoniemen: (3aS,3'R,4S,4'S,5'R,6R,6'R,7S,7aS)-4-(((1R,2S,3R,5S,6R)-3-Amino-2,6-dihydroxy-5-(methylamino)cyclohexyl)oxy)-6'-((S)-1-amino-2-hydroxyethyl)-6-(hydroxymethyl)octahydro-3aH-spiro[[1,3]dioxolo[4,5-c]pyran-2,2'-pyran]-3',4',5',7-tetraol
- CAS-nummer:31282-04-9
- Formule:C₂₀H₃₇N₃O₁₃
- MDL-nummer:MFCD06795479
- Molecular Weight:527.5 g/mol
- Merck index:14,4852
- Temperatuur bij opslag:Koelkast
- EINECS:250-545-5
- UN:3462
- ADR:6.1,I
Documents
Certificaten
Zoeken op lotnummerOrganic Building Blocks and Intermediates
Application Notes
Thermo Scientific Cell Culture Workflow
Product documentatie
Specificaties
Specificaties
- Verpakking:Glass bottle for solids
- VPE:100 mg
Specificaties
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



