Orders LinkContactUsLinkComponent
MES-SDS running buffer (20X)
MES-SDS running buffer (20X)
MES-SDS running buffer (20X), 500 mL
Catalogus # J62138.AP
Leverancier:   Thermo Fisher Scientific
MES-SDS running buffer (20X)
Catalogus # J62138.AP
CAS-nummer:  
MES-SDS running buffer (20X)
MES-SDS running buffer (20X)
MES-SDS running buffer (20X), 500 mL
CAS-nummer:  
Price
124,00 €
Log in voor beschikbaarheid
1 * 500 mL
Product Details & Documents
Physical Form: Liquid
Specificaties
Specificaties
  • VPE:
    500 mL
  • Omschrijving:
    MES-SDS running buffer (20X)
  • Cat. No.:
    J62138.AP
  • Physical form:
    Liquid
Specificaties