Buffers, MOPS, 1,0 M solutions
MOPS buffer solution pH 8 (1 mol/l), 8,0, Glass bottle, 500 mL
Leverancier: Thermo Fisher Scientific
Buffers, MOPS, 1,0 M solutions
Catalogus # J61958.AP
Leverancier: Thermo Fisher Scientific
CAS-nummer:
Ga naar:
Product Details & Documents
CAS No.: 1132-61-2
Physical form: liquid
Appearance: colourless
Odour: odourless
MDL No.: MFCD00006183
Specificaties
Specificaties
- Verpakking:Glass bottle
- VPE:500 mL
- Omschrijving:MOPS buffer solution pH 8 (1 mol/l)
- pH-waarde:8,0
- Cat. No.:J61958.AP
- Uiterlijk:Colourless
- Geur/lucht:Odourless
- Physical form:Liquid
- MDL No.:MFCD00006183
- CAS No.:1132-61-2



