Orders LinkContactUsLinkComponent
TBE sample buffer (6X)
TBE Sample Buffer, 6X, 50 mL
Catalogus # J60357.AD
Leverancier:   Thermo Fisher Scientific
TBE sample buffer (6X)
Catalogus # J60357.AD
CAS-nummer:  
TBE sample buffer (6X)
TBE sample buffer (6X)
TBE Sample Buffer, 6X, 50 mL
CAS-nummer:  
Price
75,80 €
Log in voor beschikbaarheid
1 * 50 mL
Product Details & Documents
Physical Form: Liquid
Appearance: Blue
Specificaties
Specificaties
  • VPE:
    50 mL
  • Omschrijving:
    TBE Sample Buffer, 6X
  • Cat. No.:
    J60357.AD
  • Uiterlijk:
    Blue
  • Physical form:
    Liquid