Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Buffers, MOPS, 1,0 M solutions

Buffers, MOPS, 1,0 M solutions

MOPS buffer solution pH 7,5 (1 mol/l), 7,5, Glass bottle, 500 mL
Catalogus # J61843.AP
Leverancier:   Thermo Fisher Scientific
Buffers, MOPS, 1,0 M solutions
Catalogus # J61843.AP
CAS-nummer:  
Buffers, MOPS, 1,0 M solutions
Buffers, MOPS, 1,0 M solutions

MOPS buffer solution pH 7,5 (1 mol/l), 7,5, Glass bottle, 500 mL

CAS-nummer:  
Price
223,00 €
Log in voor beschikbaarheid
1 * 500 mL
Product Details & Documents

CAS No.: 1132-61-2
Physical form: liquid
Appearance: colourless
Odour: odourless
MDL No.: MFCD00006183

Specificaties

Specificaties

  • Verpakking:
    Glass bottle
  • VPE:
    500 mL
  • Omschrijving:
    MOPS buffer solution pH 7,5 (1 mol/l)
  • pH-waarde:
    7,5
  • Cat. No.:
    J61843.AP
  • Uiterlijk:
    Colourless
  • Geur/lucht:
    Odourless
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00006183
  • CAS No.:
    1132-61-2

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.