Fast Blue BB base
100 g
Leverancier: Thermo Fisher Scientific
Ga naar:
Product Details & Documents
CAS No.: 120-00-3 Molecular Formula: C17H20N2O3 Formula Weight: 300.35 Physical Form: Powder Appearance: Gray to pale brown Melting Point: 95-105° Beilstein Reference: 3416889 MDL No.: MFCD00009091
Applications: C.I. 37175
Synoniemen: 4'-Amino-2',5'-diethoxybenzanilide, N-(4-Amino-2,5-diethoxyphenyl)benzamide, Azoic Diazo No. 20, Fast blue BB, C.I. 37175, Azoic diazo component 20 (base)
- CAS-nummer:120-00-3
- Formule:C₁₇H₂₀N₂O₃
- MDL-nummer:MFCD00009091
- Molecular Weight:300.36 g/mol
- Smelten Pt:95…105 °C
- Temperatuur bij opslag:Kamertemperatuur
- EINECS:204-363-8
Specificaties
Specificaties
- VPE:100 g



