Buffers, MOPS, 1,0 M solutions
MOPS buffer solution pH 7 (1 mol/l), 7,0, Glass bottle, 250 mL
CAS No.: 1132-61-2
Physical form: liquid
Appearance: colourless
Odour: odourless
MDL No.: MFCD00006183
- Verpakking:Glass bottle
- VPE:250 mL
- Omschrijving:MOPS buffer solution pH 7 (1 mol/l)
- pH-waarde:7,0
- Cat. No.:J61821.AK
- Uiterlijk:Colourless
- Geur/lucht:Odourless
- Physical form:Liquid
- MDL No.:MFCD00006183
- CAS No.:1132-61-2
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



