Buffers, MOPS, 1,0 M solutions
MOPS buffer solution pH 6,5 (1 mol/l), 6,5, Glass bottle, 500 mL
Leverancier: Thermo Fisher Scientific
Buffers, MOPS, 1,0 M solutions
Catalogus # J61797.AP
Leverancier: Thermo Fisher Scientific
CAS-nummer:
Ga naar:
Product Details & Documents
CAS No.: 1132-61-2
Physical form: liquid
Appearance: colourless
Odour: odourless
MDL No.: MFCD00006183
Documents
Specificaties
Specificaties
- Verpakking:Glass bottle
- VPE:500 mL
- Omschrijving:MOPS buffer solution pH 6,5 (1 mol/l)
- pH-waarde:6,5
- Cat. No.:J61797.AP
- Uiterlijk:Colourless
- Geur/lucht:Odourless
- Physical form:Liquid
- MDL No.:MFCD00006183
- CAS No.:1132-61-2
Specificaties
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



