Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Kinase buffer II (5X)

Kinase buffer II (5X)

Kinase buffer II (5X), 250 mL
N° de catalogue J60269.AK
Fournisseur:   Thermo Fisher Scientific
Kinase buffer II (5X)
N° de catalogue J60269.AK
Numéro CAS:  
Kinase buffer II (5X)
Kinase buffer II (5X)

Kinase buffer II (5X), 250 mL

Numéro CAS:  
Prix
78,80 €
Connexion Pour la disponibilité
1 * 250 mL
Product Details & Documents

Physical form: Liquid

Spécifications

Spécifications

  • VPE:
    250 mL
  • Omschrijving:
    Kinase buffer II (5X)
  • Cat. No.:
    J60269.AK
  • Physical form:
    Liquid

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.