MOPS-SDS running buffer (20X)
MOPS-SDS running buffer (20X), 1 L
Leverancier: Thermo Fisher Scientific
MOPS-SDS running buffer (20X)
Catalogus # J62847.K2
Leverancier: Thermo Fisher Scientific
CAS-nummer:
Ga naar:
Product Details & Documents
Physical Form: Liquid
Documents
Specificaties
Specificaties
- VPE:1 L
- Omschrijving:MOPS-SDS running buffer (20X)
- Cat. No.:J62847.K2
- Physical form:Liquid