TE buffer, (20X), pH 8,0, autoclaved
TE buffer concentrate, autoclaved, 20X, 8,0, 250 mL
Leverancier: Thermo Fisher Scientific
TE buffer, (20X), pH 8,0, autoclaved
Catalogus # J62388.AK
Leverancier: Thermo Fisher Scientific
CAS-nummer:
Ga naar:
Product Details & Documents
Physical Form: Liquid
MDL No.: MFCD00236359
Documents
Specificaties
Specificaties
- Concentratie:20X
- VPE:250 mL
- Omschrijving:TE buffer concentrate, autoclaved
- pH-waarde:8,0
- Cat. No.:J62388.AK
- Physical form:Liquid
- MDL No.:MFCD00236359



