Order Entry
Belgium
ContactUsLinkComponent
[EN]QUALITATIVE FLAT FILTER 125MM 6-10µm 1 * 100 ST
Catalogus # PRATA125110
Leverancier:  PRAT DUMAS
[EN]QUALITATIVE FLAT FILTER 125MM 6-10µm 1 * 100 ST
Catalogus # PRATA125110
Leverancier:  PRAT DUMAS
Frequently Bought Together