Orders LinkContactUsLinkComponent
Mortars without pestles
Mortars without pestles
100 ml, 60 mm, 80 mm
Catalog # BIBB718234
Supplier:   SciLabware
Mortars without pestles
Catalog # BIBB718234
Supplier:  SciLabware
Mortars without pestles
Mortars without pestles
100 ml, 60 mm, 80 mm
Price
57,10 €
Log in for availability
1 * 1 ST
Product Details & Documents
Ideal for mixing and grinding powders.
  • Borosilicate glass
  • Low form model
Ordering information: Please order pestles separately.
Specifications
  • Ø:
    80 mm
  • Capaciteit:
    100 ml
  • Hoogte:
    60 mm