Statieven
Head holder for Ø 25 mm pole and Ø 76 mm head with mount rack and focusing device
Catalogus # MOTI1101010100242
Leverancier: MOTIC
Ga naar:
Product Details & Documents
Services: Installatie, IQOQ, onderhoud of reparatie nodig? Mail naar [email protected]
Specificaties
- Omschrijving:Head holder for Ø 25 mm pole and Ø 76 mm head with mount rack and focusing device
- Seq:13