Disposable cannulas
Disposable cannulas, G25, oranje, 60 mm, +, 0,5×60 mm
Catalog # EHRS15884260
Supplier: EHRHARDT SOEHNE
Jump to:
Product Details & Documents
Disposable cannulas.
- Pyrogen-free, not toxic
- With lancet cut
- EO-sterilisation
Documents
Certificates
Search by Lot NumberSpecifications
- Ø×L:0,5×60 mm
- Meter:G25
- Steriel:+
- Kleur:oranje
- Omschrijving:Disposable cannulas
- Lengte:60 mm