0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Belgium
ContactUsLinkComponent
Trans-blot turbo transfer pack,midi nitrocellulose,pkg of 10 1 * 10 ST
BRAD170-4159
:
BIO RAD
Trans-blot turbo transfer pack,midi nitrocellulose,pkg of 10 1 * 10 ST
BRAD170-4159
:
BIO RAD
Frequently Bought Together
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
ep Dualfilter T.I.P.S.® Racks, Filter Pipette Tips
: EPPENDORF
VWR® Standaardlijntransferpipetten
: VWR Collection
VWR®, Transferpipetten, Hoge Prestaties
: VWR Collection
VWR®, Verspreiders, L-vormig, Wegwerpbaar
: VWR Collection
VWR®, Rechte monsterrecipiënt met schroefdop
: VWR Collection
VWR® PCR-buisjes, 0,2 ml
: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
: Corning
VWR® Inoculating Loops and Needles, Disposable
: VWR Collection
VWR®, Serologische pipetten, standaardreeks
: VWR Collection
VWR®, Centrifugeerbuisjes, conische bodem
: VWR Collection
GOAT ANTI-RABBIT IMMUNOGLOBULINS/HRP 1 * 1 mL
: Dako