Orders LinkContactUsLinkComponent
Krukken
Stool 2, black, made from stamskin top synthetic leather upholstery, 460 - 630 mm, Castors, —
Catalogus # BIMO9468H-2571-807
Leverancier:   bimos
Krukken
Catalogus # BIMO9468H-2571-807
Leverancier:  bimos
Krukken
Krukken
Stool 2, black, made from stamskin top synthetic leather upholstery, 460 - 630 mm, Castors, —
Price
163,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Production stools with Extra large 40 cm Ø seats.
  • Hardwearing stools provide optimum support in industrial and workshop applications
  • Sturdy steel 5 star base
  • Encourage the posture in keeping with the natural S-shape of the spine
  • Seat height adjustment based on pneumatic spring system
ESD standing rest and ESD stools:
Pneumatic spring height adjustment system
With conductive fabric or synthetic leather upholstery
Conductive castors or glides (stools)
Cleanroom stools:
Base frame made from die-cast aluminium, brilliantly polished
Conductive castors/glides for hard floors
Synthetic leather cover with conductive and non-slip properties
Laboratory Stools:
The new stool system from Bimos is a comfortable and flexible solution for short periods of sitting
Its configuration options make the stool system a versatile seating solution for industrial applications, the service sector, healthcare settings, as well as laboratories
Easy ring control
Coloured ring in various colours
Polished aluminium base
Resistant to desinfectants
Specificaties
  • Omschrijving:
    Stool 2, black, made from stamskin top synthetic leather upholstery
  • Hoogte:
    460 - 630 mm
  • Seq:
    7
  • Voetvorm:
    Castors
  • Voetring:
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.