(2-Hydroxylpropyl)-β-cyclodextrine
Glass bottle, 100 g
Catalogus # TCIAH0979-100G
Leverancier: TCI
(2-Hydroxylpropyl)-β-cyclodextrine
Catalogus # TCIAH0979-100G
Leverancier: TCI
CAS-nummer: 128446-35-5
Ga naar:
Product Details & Documents
Synoniemen: Hydroxypropyl Betadex, Hydroxypropyl-β-cyclodextrin, (2-Hydroxypropyl)-β-cyclodextrin, HP-β-CD
- CAS-nummer:128446-35-5
- Formule:C₆₃H₁₁₂O₄₂
- MDL-nummer:MFCD00074978
- Temperatuur bij opslag:Kamertemperatuur
- EINECS:420-920-1
Specificaties
Specificatie testresultaten
- Appearance:White to Almost white powder to crystal
- Specific rotation [a]20/D:+120 to +135 deg (C=1, H2O)
- Drying loss:max. 10.0 %
Specificaties
- Verpakking:Glass bottle
- VPE:100 g
Specificaties
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



