Orders LinkContactUsLinkComponent
Eggertz tubes for comparators
Eggertz tubes for comparators
Eggertz tubes, 20 ml with 0,1 ml division for comparators
Catalogus # WITG2.149.020
Leverancier:   witeg Labortechnik
Eggertz tubes for comparators
Catalogus # WITG2.149.020
Leverancier:  witeg Labortechnik
Eggertz tubes for comparators
Eggertz tubes for comparators
Eggertz tubes, 20 ml with 0,1 ml division for comparators
Price
42,80 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Specificaties
  • Omschrijving:
    Eggertz tubes, 20 ml with 0,1 ml division for comparators
  • Seq:
    01