Sample containers, straight, with screw cap, WHEATON®
100 ml, Plain label, sterile, helder, PS / metaal
Catalogus # 216-1012
Leverancier: DWK Life Sciences
Sample containers, straight, with screw cap, WHEATON®
Catalogus # 216-1012
Leverancier: DWK Life Sciences
Ga naar:
Product Details & Documents
Polystyrene clear, with metal screw cap.
- Manufactured from FDA approved virgin polystyrene
- Supplied sterile and for in vitro use only
- CE marked in accordance with European Directive 98/79/EC
- All containers are leak tested to British Standard BS5213:1975(1989)
- All materials are non cytotoxic
Straight walled vessels ideal for sample collection, storage and transport. Polystyrene offers good thermal and chemical resistance. Supplied with flow seal metal cap.
Let op: *De product(en) op deze pagina vallen buiten het toepassingsgebied van de EU-richtlijn 2019/904
Documents
Specificaties
- Type:Plain label, sterile
- Capaciteit:100 ml
- Kleur:helder
- Materiaal:PS / metaal