Sample containers, with screw cap, universal, WHEATON®
30 ml, Universal container, plain label, sterile, helder, PS/PE
Leverancier: DWK Life Sciences
Sample containers, with screw cap, universal, WHEATON®
Catalogus # 216-0982
Leverancier: DWK Life Sciences
Ga naar:
Product Details & Documents
PP translucent or PS clear, with PE screw cap.
- Manufactured from FDA approved virgin polystyrene or polypropylene
- PS offers good thermal and chemical resistance
- Supplied sterile - for in vitro use only
- All materials are non cytotoxic
Straight walled containers ideal for sample collection, storage and transport. Supplied with leakproof, flow seal closure. Available pre-filled with boric acid for urine samples.
Certificeringen: CE marked in accordance with European Directive 98/79/EC
All containers are leak tested to British Standard BS5213:1975(1989)
Verpakking: Packed in inner bags of 50.
Let op: *De product(en) op deze pagina vallen buiten het toepassingsgebied van de EU-richtlijn 2019/904
Documents
Specificaties
- Type:Universal container, plain label, sterile
- Capaciteit:30 ml
- Kleur:helder
- Materiaal:PS/PE