Celtellers, Neubauer
Counting chamber, bright-lined, Neubauer, 0,10 mm
Catalogus # MARI0640130
Leverancier: MARIENFELD
Ga naar:
Product Details & Documents
These counting chambers are designed for counting leucocytes and erythrocytes.
- Double net ruling
- Two counting grids with one division
Certificeringen: IVD 98/79/EC
Levering: Supplied with two cover glasses.
Documents
Certificaten
Zoeken op lotnummerSpecificaties
- Kamerdiepte:0,10 mm
- Omschrijving:Counting chamber, bright-lined, Neubauer
- Seq:02