Inlaat liners
Liner, ultra inert, splitless, singletaper, no wool, 4 mm
HEWL5190-3162
: Agilent
These inlet liners prevent adsorption and ensure accurate sample transfer onto the GC column through a robust deactivated surface. These liners are recommended for trace level analysis of active analytes. Liners with inert wool retain non-volatiles, prolonging column life and lowering frequency of MS source maintenance.
- Consistent high level of inertness
- Ø inw.:4 mm
- Omschrijving:Liner, ultra inert, splitless, singletaper, no wool
- Seq:26