Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
QuEChERS dispersive kits
Buisje met 50 mg primaire/secundaire amine, 150 mg magnesiumsulfaat: met keramische homogenisatoren, verwijdert polaire organische zuren, sommige suikers en lipiden, 2 ml
  HEWL5982-5022CH
 :   Agilent
QuEChERS dispersive kits
  HEWL5982-5022CH
 :  Agilent
QuEChERS dispersive kits
QuEChERS dispersive kits
Buisje met 50 mg primaire/secundaire amine, 150 mg magnesiumsulfaat: met keramische homogenisatoren, verwijdert polaire organische zuren, sommige suikers en lipiden, 2 ml
188,00 €
1 * 1 ST
This product has been blocked by your organization. Please contact your purchasing department for more information
Step 2: Dispersive SPE cleanup
Select the dispersive SPE kit suited to the type of food being analysed and the method you are following. In this step, an aliquot of the sample extract from step one (see QuEChERS extraction kits) is added to a 2 or 15 ml centrifuge tube containing a small amount of SPE sorbent and MgSO₄. The sorbent will pull out interfering matrix materials from the sample, while the MgSO₄ helps remove excess water and improve analyte partitioning.
  • Capaciteit:
    2 ml
  • Omschrijving:
    Buisje met 50 mg primaire/secundaire amine, 150 mg magnesiumsulfaat: met keramische homogenisatoren, verwijdert polaire organische zuren, sommige suikers en lipiden
  • Seq:
    02