α-Cyclodextrin ≥98.0% (by HPLC)
Glass bottle, 10 g
Catalogus # TCIAC0776-10G
Leverancier: TCI
Ga naar:
Product Details & Documents
- CAS-nummer:10016-20-3
- Formule:C₃₆H₆₀O₃₀
- MDL-nummer:MFCD00078207
- Molecular Weight:972.84 g/mol
- Merck index:14,02718
- EINECS:233-007-4
Specificaties
Specificatie testresultaten
- Appearance:White powder to crystal
- Purity (HPLC):min. 98.0 area%
- Specific rotation [a]20/D:+146.0 to +153.0 deg (C=1, H2O) (calcd.on anh.substance)
- Solubility in Water:almost transparency
- Water:max. 10.0 %
Specificaties
- Verpakking:Glass bottle
- VPE:10 g



