Test panels
Test panel, cold rolled steel, smooth, degreased and cleaned, 60×200×0,5 mm
Catalogus # SIMEVF8510
Leverancier: Industrial Physics
Ga naar:
Product Details & Documents
Test panels are used to enhance reproducibility of physical and chemical tests. Each panel is equipped with a hole for hanging and handling.
- Consistent and reproducible quality with sophisticated finishing: No burrs or sharp edges
- Physical testing: Adhesion, elongation, impact resistance and elasticity
- Appearance testing of coatings (gloss, colour, texture)
Documents
TQC Company Video
Video 's
Specificaties
- Omschrijving:Test panel, cold rolled steel, smooth, degreased and cleaned
- Seq:16
- b×d×h:60×200×0,5 mm
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.