Test panels
Test panel, cold rolled steel, smooth, degreased and cleaned, 75×150×0,5 mm
Catalogus # SIMEVF8511
Leverancier: Industrial Physics
Ga naar:
Product Details & Documents
Test panels are used to enhance reproducibility of physical and chemical tests. Each panel is equipped with a hole for hanging and handling.
- Consistent and reproducible quality with sophisticated finishing: No burrs or sharp edges
- Physical testing: Adhesion, elongation, impact resistance and elasticity
- Appearance testing of coatings (gloss, colour, texture)
Documents
TQC Company Video
Video 's
Specificaties
- Omschrijving:Test panel, cold rolled steel, smooth, degreased and cleaned
- Seq:17
- b×d×h:75×150×0,5 mm