Orders LinkContactUsLinkComponent
Impact testers
Impact testers
Dual impact tester, ISO 6272-1 and ASTM D2794 (before 1993), incl. base plate assembly, guide tube, release collar, clamp device, ball Ø 20 mm, die 27 mm, 1 kg, ball Ø 15,9 mm and die 16,3 mm, 0,9 kg
Catalogus # SIMESP1891
Leverancier:   Industrial Physics
Impact testers
Catalogus # SIMESP1891
Leverancier:  Industrial Physics
Impact testers
Impact testers
Dual impact tester, ISO 6272-1 and ASTM D2794 (before 1993), incl. base plate assembly, guide tube, release collar, clamp device, ball Ø 20 mm, die 27 mm, 1 kg, ball Ø 15,9 mm and die 16,3 mm, 0,9 kg
Price
2 630,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Impact testers used to determine the impact resistivity and flexibility of materials.
  • Spirit level built-in for correct positioning
  • Dual scale instrument equipped with a special guidance
  • Distance between each impact is always according to the standards
Certificeringen: DIN 6272-1 and -2, ASTM D2794 and G14.
Documents
Specificaties
  • Omschrijving:
    Dual impact tester, ISO 6272-1 and ASTM D2794 (before 1993), incl. base plate assembly, guide tube, release collar, clamp device, ball Ø 20 mm, die 27 mm, 1 kg, ball Ø 15,9 mm and die 16,3 mm, 0,9 kg
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.