Test panels
Test panel, aluminium, matt, degreased and cleaned, 50×100×0,8 mm
Catalogus # SIMEVF8522
Leverancier: Industrial Physics
Ga naar:
Product Details & Documents
Test panels are used to enhance reproducibility of physical and chemical tests. Each panel is equipped with a hole for hanging and handling.
- Consistent and reproducible quality with sophisticated finishing: No burrs or sharp edges
- Physical testing: Adhesion, elongation, impact resistance and elasticity
- Appearance testing of coatings (gloss, colour, texture)
Documents
TQC Company Video
Video 's
Specificaties
- Omschrijving:Test panel, aluminium, matt, degreased and cleaned
- Seq:01
- b×d×h:50×100×0,8 mm