Orders LinkContactUsLinkComponent
Test panels
Test panels
Test panel, aluminium, matt, degreased and cleaned, 60×140×0,6 mm
Catalogus # SIMEVF8530
Leverancier:   Industrial Physics
Test panels
Catalogus # SIMEVF8530
Leverancier:  Industrial Physics
Test panels
Test panels
Test panel, aluminium, matt, degreased and cleaned, 60×140×0,6 mm
Price
560,00 €
Log in voor beschikbaarheid
1 * 600 ST
Product Details & Documents
Test panels are used to enhance reproducibility of physical and chemical tests. Each panel is equipped with a hole for hanging and handling.
  • Consistent and reproducible quality with sophisticated finishing: No burrs or sharp edges
  • Physical testing: Adhesion, elongation, impact resistance and elasticity
  • Appearance testing of coatings (gloss, colour, texture)
Documents
Specificaties
  • Omschrijving:
    Test panel, aluminium, matt, degreased and cleaned
  • Seq:
    02
  • b×d×h:
    60×140×0,6 mm