Orders LinkContactUsLinkComponent
Inspection sets
Inspection set with destructive wood moisture meter
Catalogus # SIMESP5000
Leverancier:   Industrial Physics
Inspection sets
Catalogus # SIMESP5000
Leverancier:  Industrial Physics
Inspection sets
Inspection sets
Inspection set with destructive wood moisture meter
Price
756,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
These basic sets are designed to perform all measurements and tests regarding painting.
Delivery information: Moisture meter, digital thermo/hygrometer, compass, illuminated surface microscope (to check the surface and paint layer), stainless steel cutter knife, pH-indicator paper (to check the acidity of concrete walls and floors), wedge (for checking reverse-side tolerance and thickness of glass), telescopic inspection mirror and lamp (for visual control), waterproof marker and MasterPaintPlate (multifunctional instrument to measure wet film thickness, cover of edges, thickness of glass, as well as providing an indication of the viscosity and adhesion of the coating).
Documents
Specificaties
  • Omschrijving:
    Inspection set with destructive wood moisture meter
  • Seq:
    001
Accessories
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • Knife with rubber grip
    Knife with rubber grip
    SIMESP3008
    Log in voor beschikbaarheid
    1 * 1 ST - 30,50 €
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • Sturdy stainless steel bradle for dry rot inspection in timber
    Sturdy stainless steel bradle for dry rot inspection in timber
    SIMESP6202
    Log in voor beschikbaarheid
    1 * 1 ST - 74,40 €
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • Water resistant marker
    Water resistant marker
    SIMESP6400
    Log in voor beschikbaarheid
    1 * 1 ST - 13,50 €