Copper(I) bromide ≥98%
50 g
Purity: 98% CAS No.: 7787-70-4 Molecular Formula: BrCu Formula Weight: 143.45 Physical Form: Powder Density: 4.98 Appearance: Pale green Melting Point: 492° Boiling Point: 1345° Solubility: Slightly soluble in cold water. Decomposes in hot water. Soluble in HBr, HCl, NH4OH Merck Reference: 14,2659 UN No.: 3077 MDL No.: MFCD00010969
Catalyst
- :7787-70-4
- :BrCu
- :MFCD00010969
- :143.45 g/mol
- :14,2659
- :1345 °C
- :492 °C
- :4,98
- :Kamertemperatuur
- :232-131-6
- :9,III
- :Density: 4,71 g/cm³ (20 °C)
- VPE:50 g