Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Copper(I) bromide ≥98%
50 g
  11605.18
Copper(I) bromide ≥98%
  11605.18
 :  7787-70-4
Copper(I) bromide ≥98%
Copper(I) bromide ≥98%
50 g
 :  7787-70-4
28,50 €
1 * 50 g
Purity: 98% CAS No.: 7787-70-4 Molecular Formula: BrCu Formula Weight: 143.45 Physical Form: Powder Density: 4.98 Appearance: Pale green Melting Point: 492° Boiling Point: 1345° Solubility: Slightly soluble in cold water. Decomposes in hot water. Soluble in HBr, HCl, NH4OH Merck Reference: 14,2659 UN No.: 3077 MDL No.: MFCD00010969
Catalyst
  •  :7787-70-4
  •  :BrCu
  •  :MFCD00010969
  •  :143.45 g/mol
  •  :14,2659
  •  :1345 °C
  •  :492 °C
  •  :4,98
  •  :Kamertemperatuur
  •  :232-131-6
  •  :9,III
  •  :Density: 4,71 g/cm³ (20 °C)
  • VPE:
    50 g