Inlaat liners
Split/splitless fast FocusLiner, with fused silica wool, 2,3 mm
Catalogus # HEWLSG092005
Leverancier: Agilent
Ga naar:
Product Details & Documents
These inlet liners prevent adsorption and ensure accurate sample transfer onto the GC column through a robust deactivated surface. These liners are recommended for trace level analysis of active analytes. Liners with inert wool retain non-volatiles, prolonging column life and lowering frequency of MS source maintenance.
- Consistent high level of inertness
Specificaties
- Ø inw.:2,3 mm
- Omschrijving:Split/splitless fast FocusLiner, with fused silica wool
- Seq:27