Orders LinkContactUsLinkComponent
pH buffer solutions
pH buffer standard solution with certificate of analysis, 125 ml, 10,00, 1
Catalogus # HACHLZW9470.99
Leverancier:   Hach
pH buffer solutions
Catalogus # HACHLZW9470.99
Leverancier:  Hach
CAS-nummer:  
pH buffer solutions
pH buffer solutions
pH buffer standard solution with certificate of analysis, 125 ml, 10,00, 1
CAS-nummer:  
Price
16,40 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Specificaties
Specificaties
  • VPE:
    1
  • Omschrijving:
    pH buffer standard solution with certificate of analysis, 125 ml
  • pH-waarde:
    10,00
Specificaties