Orders LinkContactUsLinkComponent
Gel drying Frames™
Gel drying frame, 140×140 mm
  DIVEAB-110760
Gel drying Frames™
  DIVEAB-110760
Gel drying Frames™
Gel drying Frames™
Gel drying frame, 140×140 mm
363,00 €
1 * 1 KIT
Gel drying frames allow researchers for easy handling and crack free dry gels without the use of heat/vacuum dryers.
 : The complete kit includes 2 open frames, 2 solid back plates, 12 side clips and 50 sheets of pre-cut cellophane.
  • Omschrijving:
    Gel drying frame, 140×140 mm
  • Seq:
    01