Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Safety helmets, Concept

Safety helmets, Concept

Safety helmet, vented, without ratchet, EU, rood
Catalogus # 111-0866
Safety helmets, Concept
Catalogus # 111-0866
Safety helmets, Concept
Safety helmets, Concept

Safety helmet, vented, without ratchet, EU, rood

Price
25,30 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents

Lightweight, comfortable and modern protective helmet made from ABS (acrylonitrile butadiene styrene) with large forehead area featuring an imprint for the company logo.

  • With extended nape area
  • Six point terylene inner cradle
  • Highly absorbent hydro-flock sweatband
  • With increased side rigidity (additional lateral deformity test)
  • Wide ventilation slots
  • Without ratchet, with full peak

Size: 510 to 630 mm

Weight: 325 g

EN 397

Certificeringen: EN 397

Documents

Specificaties
  • Kleur:
    rood
  • Omschrijving:
    Safety helmet, vented, without ratchet, EU
  • Seq:
    04
Aanbevelingen

Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.

Anders ontvangt u algemene aanbevelingen.