Arecolinium bromide
Glass bottle for solids, 250 mg
Catalogus # CAYM13662-250
Leverancier: Cayman Chemical
Ga naar:
Product Details & Documents
Synoniemen: Arecoline hydrobromide
- CAS-nummer:300-08-3
- Formule:C₈H₁₄BrNO₂
- MDL-nummer:MFCD00039041
- Molecular Weight:236.11 g/mol
- EINECS:206-087-3
- UN:1544
- ADR:6.1,III
Specificaties
Specificaties
- Verpakking:Glass bottle for solids
- VPE:250 mg



